Learn More
Medchemexpress LLC Verapamil EP Impurity C hydrochloride | 51012-67-0 | 99.6% | 245.75 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13658910MM/1ML
Encompass_Preferred
Verapamil EP Impurity C hydrochloride, also known as NSC-609249 hydrochloride, is an impurity of Verapamil. Verapamil itself acts as a calcium channel blocker and a potent, orally active first-generation P-glycoprotein (P-gp) inhibitor. This product is intended for research use only.
  • Impurity of Verapamil
  • Related to a calcium channel blocker
  • Related to a P-glycoprotein inhibitor
  • For research use only
Catalog No. 50-003-51939