Learn More
U.S. Pharmacopeia Verapamil hydrochloride, 152-11-4, MFCD00055208, 200 mg
Supplier:  U.S. Pharmacopeia 1711202
Encompass
Linear Formula: (CH3O)2C6H3CH2CH2N(CH3)(CH2)3C[C6H3(OCH3)2][CH(CH3)2]CN · HCl, Molecular Weight: 491.06, mp: 142 °C, Synonym: (±)-Verapamil hydrochloride, 5-[N-(3,4-Dimethoxyphenylethyl)methylamino]-2-(3,4-dimethoxyphenyl)-2-isopropylvaleronitrile hydrochloride.
Catalog No. NC1343858