Learn More
Medchemexpress LLC Verapamil hydrochloride | 152-11-4 | MFCD00055208 | 99.8% | 491.06 g/mol | C27H39ClN2O4 | 10 MG

Supplier: Medchemexpress LLC HYA0064R10MG
Verapamil hydrochloride is supplied as a high-purity analytical standard of the verapamil hydrochloride salt intended for laboratory research and analytical applications. It is commonly used in pharmacology and transport studies involving calcium channel activity and P-glycoprotein.
- High purity: 99.8% analytical grade
- Suitable as an analytical and pharmacological standard
- Solid form supplied in a 10 mg quantity
- Molecular weight approximately 491.06 g/mol
- CAS number: 152-11-4 for unambiguous identification
- Certificate of analysis and safety data sheet available
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.