Learn More
Abcam Verapamil hydrochloride (mM/ml), L-type Ca2+ channel blocker, 50MM_ML
Supplier:  Abcam AB14668050MM_ML
Encompass_Preferred
MW 491.1 Da, Purity >99%. L-type Ca2+ channel blocker. Antiarrhythmic in vivo. Soluble in 1 ml water to give specified mM/ml concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4529