Learn More
Medchemexpress LLC Verapamil hydrochloride (Standard) | 152-11-4 | 99.8% | C27H39ClN2O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYA0064R50MG
Encompass_Preferred
Verapamil hydrochloride (Standard) is the analytical standard of Verapamil hydrochloride, intended for research and analytical applications. It is a calcium channel blocker and a potent, orally active first-generation P-glycoprotein (P-gp) inhibitor, also inhibiting CYP3A4. It has potential for high blood pressure, heart arrhythmias, and angina research.
  • Used as an analytical standard and reference standard in assays
  • Commonly employed in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS
Catalog No. 50-004-32983