Learn More
U.S. Pharmacopeia Verapamil Related Compound F, 93-03-8, MFCD00004638, 150 mg
Supplier:  U.S. Pharmacopeia 1711440
Encompass
Linear Formula: (CH3O)2C6H3CH2OH, Molecular Weight: 168.19, bp: 296-297 °C, Synonym: 3,4-Dimethoxybenzyl alcohol, Veratryl alcohol.
Catalog No. NC1355528