Learn More
NEOGEN Veratox for DON 2/3, 38 tests
SDP
Supplier:  NEOGEN 700002534
Encompass_Preferred
Veratox for DON 2/3 is a competitive direct ELISA that provides a quantitative analysis of DON in cereal grains and grain products, such as wheat, wheat flour, wheat midds, wheat bran, corn, cornmeal, corn screenings, barley, malted barley and oats.
Catalog No. NC1835033