Learn More
NEOGEN Veratox for Zearalenone
SDP
Supplier:  NEOGEN 700002513
Encompass_Preferred
Veratox for Zearalenone is a competitive direct ELISA that provides a quantitative analysis of zearalenone in such commodities as corn, wheat, barley and DDGS.
Catalog No. NC1835032