Learn More
Medchemexpress LLC Veratraldehyde | 120-14-9 | 166.18 | 1 KG
SDP
Supplier:  Medchemexpress LLC HYN10961KG
Encompass_Preferred
Veratraldehyde is an orally active aromatic compound that exhibits antibacterial properties. It can be naturally isolated from the essential oils of plants such as peppermint and ginger. This compound functions as an antibacterial agent by targeting the PilY1 protein and has demonstrated activity against *Pseudomonas aeruginosa*. Additionally, Veratraldehyde is noted for its repellent effects against mosquitoes and ticks and can also be utilized as a flavoring agent.
  • Orally active aromatic compound
  • Natural origin: isolated from plant essential oils like peppermint and ginger
  • Antibacterial agent: effectively targets the PilY1 protein to inhibit bacterial growth
  • Activity against *Pseudomonas aeruginosa*
  • Repellent effect: acts as a repellent against mosquitoes and ticks
  • Flavoring agent
Catalog No. 50-004-15590