Learn More
Medchemexpress LLC Veratraldehyde | 120-14-9 | 166.18 | 100 G

Supplier: Medchemexpress LLC HYN1096100G
Veratraldehyde is an orally active aromatic compound and an antibacterial agent. It can be isolated from essential oils of plants such as peppermint and ginger. This compound targets the PilY1 protein and exhibits antibacterial activity against Pseudomonas aeruginosa. Additionally, Veratraldehyde has a repellent effect against mosquitoes and ticks. It can also be used as a flavoring agent.
- Purity: 99.94%
- Chemical formula: C9H10O3
- Appearance: Solid
- Color: White to light yellow
- Initial source: Plants (Scrophulariaceae Lagotis alutacea W. W. Smith)
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage: 4°C, stored under nitrogen. In solvent: -80°C for 6 months; -20°C for 1 month (stored under nitrogen).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.