Learn More
Medchemexpress LLC HY-N2007 100mg Medchemexpress, Veratric acid CAS:93-07-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN2007100MG
Encompass_Preferred
Medchemexpress, HY-N2007 100mg Veratric acid CAS:93-07-2 Veratric acid (3,4-Dimethoxybenzoic acid) is an orally active phenolic compound derived from vegetables and fruits, has antioxidant [1] and anti-inflammatory activities [3] . Veratric acid also acts as a protective agent against hypertension-associated cardiovascular remodelling [2] . Veratric acid reduces upregulated COX-2 expression, and levels of PGE2, IL-6 after UVB irradiation [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3869