Learn More
Medchemexpress LLC Veratric acid (3,4-dimethoxybenzoic acid) | 93-07-2 | 182.18 | 50 G
SDP
Supplier:  Medchemexpress LLC HYN200750G
Encompass_Preferred
Veratric acid (3,4-Dimethoxybenzoic acid) is an orally active phenolic compound derived from vegetables and fruits. It possesses antioxidant and anti-inflammatory activities and acts as a protective agent against hypertension-associated cardiovascular remodelling. It reduces upregulated COX-2 expression, and levels of PGE2, IL-6 after UVB irradiation.
  • Orally active phenolic compound
  • Derived from vegetables and fruits
  • Antioxidant and anti-inflammatory activities
  • Protects against hypertension-associated cardiovascular remodelling
  • Reduces upregulated COX-2 expression
Catalog No. 50-004-15998