Learn More
Medchemexpress LLC Veratric acid | 93-07-2 | MFCD00002500 | 182.18 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN2007R50MG
Encompass_Preferred
Veratric acid (Standard) is an analytical standard intended for research and analytical applications. This orally active phenolic compound is derived from vegetables and fruits, possessing antioxidant and anti-inflammatory activities. It also protects against hypertension-associated cardiovascular remodelling and reduces upregulated COX-2 expression, and levels of PGE2, IL-6 after UVB irradiation.
  • Analytical standard for research and analytical applications.
  • Orally active phenolic compound.
  • Derived from vegetables and fruits.
  • Possesses antioxidant and anti-inflammatory activities.
  • Protects against hypertension-associated cardiovascular remodelling.
  • Reduces upregulated COX-2 expression, and levels of PGE2, IL-6 after UVB irradiation.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-15885