Learn More
Medchemexpress LLC Veratric acid (Standard) | 93-07-2 | 182.18 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN2007R100MG
Encompass_Preferred
Veratric acid (Standard) is the analytical standard of Veratric acid. It is intended for research and analytical applications. Veratric acid (3,4-Dimethoxybenzoic acid) is an orally active phenolic compound derived from vegetables and fruits, possessing antioxidant and anti-inflammatory activities. It also acts as a protective agent against hypertension-associated cardiovascular remodelling. Veratric acid reduces upregulated COX-2 expression, and levels of PGE2, IL-6 after UVB irradiation.
  • Analytical standard for Veratric acid
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
  • Orally active phenolic compound
  • Antioxidant and anti-inflammatory activities
  • Protective agent against hypertension-associated cardiovascular remodelling
  • Reduces upregulated COX-2 expression, and levels of PGE2, IL-6 after UVB irradiation
  • Purity: 99.99%
Catalog No. 50-004-15997