Learn More
Medchemexpress LLC Veratric acid (Standard) | 93-07-2 | 182.18 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN2007R
Encompass_Preferred
Veratric acid (Standard) is the analytical standard of Veratric acid. This product is intended for research and analytical applications. Veratric acid (3,4-Dimethoxybenzoic acid) is an orally active phenolic compound derived from vegetables and fruits, with antioxidant and anti-inflammatory activities. It also acts as a protective agent against hypertension-associated cardiovascular remodelling.
  • Reduces upregulated COX-2 expression, and levels of PGE2, IL-6 after UVB irradiation.
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Reference standard supplied assay.
Catalog No. 50-003-90003