Learn More
Medchemexpress LLC Veratrole (1,2-dimethoxybenzene) | 91-16-7 | 99.5% | 1 KG
Supplier: Medchemexpress LLC HYB18121KG
Veratrole (1,2-Dimethoxybenzene) is a key compound widely found in plants that attracts pollinators. Its release exhibits a circadian rhythm, playing a significant role in plant reproduction, species differentiation, and interactions with pollinators. Additionally, Veratrole can be demethylated by cytochrome P-450 in Streptomyces setonii.
- Exists widely in plants.
- Attracts pollinators.
- Plays a role in plant reproduction, species differentiation, and interactions with pollinators.
- Can be demethylated by cytochrome P-450 in Streptomyces setonii.
- No safety issues regarding genotoxicity, repeated-dose toxicity, reproductive toxicity, skin sensitization, phototoxicity/photoallergenicity, and local respiratory toxicity at current use levels.
- Poses no risk to the aquatic environment.