Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Veratrole | 91-16-7 | 99.5% | 500 G
SDP

Supplier:  Medchemexpress LLC HYB1812500G

Encompass_Preferred

Veratrole (1,2-Dimethoxybenzene) is a compound found in plants that attracts pollinators, playing a vital role in plant reproduction and interactions. It can also be demethylated by cytochrome P-450 in *Streptomyces setonii*. This product is for research use only.

  • Molecular weight: 138.17
  • Formula: C8H10O2
  • Appearance: solid below 15°C, liquid above 15°C
  • Color: colorless to light yellow
  • SMILES: COC1=CC=CC=C1OC
  • Structure classification: others
  • Initial source: plants including Leguminosae Glycine soya, Leguminosae Cyamopsis tetragonoloba (L.) Taub., and Compositae
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage: store at room temperature for 3 years; in solvent: -80°C for 2 years, -20°C for 1 year

Catalog No. 50-004-11411


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.