Learn More
Medchemexpress LLC Veratrole-d2 | 203645-56-1 | 99.7% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1812S15MG
Encompass_Preferred
Veratrole-d2 is the deuterium-labeled Veratrole (1,2-Dimethoxybenzene), a key compound found in plants that attracts pollinators. Its release has a circadian rhythm and plays an important role in plant reproduction, species differentiation, and interactions with pollinators. Veratrole can also be demethylated by cytochrome P-450 in *Streptomyces setonii*. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules as tracers for quantitation during drug development, with deuteration potentially impacting pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled Veratrole
  • Plays a role in plant reproduction, species differentiation, and interactions with pollinators
  • Can be demethylated by cytochrome P-450
  • Used as a tracer for quantitation in drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-12895