Learn More
Medchemexpress LLC HY-107858 100mg Medchemexpress, Veratryl alcohol CAS:93-03-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY107858100MG
Encompass_Preferred
Medchemexpress, HY-107858 100mg Veratryl alcohol CAS:93-03-8 Veratryl alcohol (3,4-Dimethoxybenzenemethanol), a secondary metabolite of some lignin degrading fungi, is commonly used nonphenolic substrate for assaying ligninolytic activity [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2304