Learn More
Medchemexpress LLC Veratryl alcohol 5g | 93-03-8 | 5G
SDP
Supplier:  Medchemexpress LLC HY1078585G
Encompass_Preferred
Veratryl alcohol (3 4-Dimethoxybenzyl alcohol) is a secondary metabolite of lignin-degrading fungi commonly used as a substrate for lignin peroxidase (LiP) to measure lignin degradation activity Veratryl alcohol protects LiP from inactivation by H2O2 and prevents the accumulation of LiP III compounds Veratryl alcohol also acts as a stabilizer for manganese-dependent peroxidases (MnP) Veratryl alcohol is a quorum-sensing inhibitor (QSI) and exhibits antibacterial efficacy[1 [2 [3 [4
Catalog No. 50-002-42281