Learn More
Labelmaster Inc Vermiculite Grade 4a, Extra-Coarse, 4-Cu. Ft. Bag
SDP
Supplier:  Labelmaster Inc KTV400R
Encompass_Preferred
Grade 4A Extra-Coarse Vermiculite is a great choice when considering the need for an inert cushioning or absorbent material in hazardous material packaging. This loose absorbent is ideal for use with UN wooden boxes and Variation Two shippers.
Catalog No. NC0430369