Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.4390 in Diameter, 2 in Long, 4 Micro
Supplier:  MSC 88843909
Encompass
This Class X Minus Pin Gage features a 0.4390 in diameter and a 2 in length with a 4 Micro coating. Constructed from tool steel, this gage is suitable for the calibration of measuring equipment or for direct gaging of parts.
  • Includes certificate of accuracy with NIST traceability.
  • Hassle-free guarantee allows for exchange or refund if the product does not meet expectations, with returns accepted within 30 days of shipment in its original packaging (excludes hazardous materials and cut-to-length products).
Catalog No. 50-005-31670