Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.1280 in Diameter, 2 in Length, Black Oxide
Supplier: MSC 57260283
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. Featuring a durable black oxide finish, it offers a 0.1280 inch diameter and a 2 inch overall length. It is ideal for calibrating various measuring instruments and performing no-go inspections to ensure accuracy.
- Black oxide finish.
- Includes certificate of accuracy with NIST traceability.
- Made from steel for durability.
- Suitable for setting micrometers, verniers, and other measuring devices.
- Useful for no-go inspection.