Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.8330 in, Steel
Supplier: MSC 78380292
This Class ZZ Minus Plug & Pin Gage is precisely engineered with a 0.8330 inch diameter and a 2 inch overall length. Constructed from durable steel, it features a 10 Micro coating for enhanced performance. This gage includes a Certificate of Accuracy with NIST Traceability, making it ideal for various metrology tasks.
- Designed for setting micrometers
- Suitable for NoGo inspection
- Compatible with verniers and other measuring devices
- Features a 0.000200 inch tolerance
- Includes certificate of accuracy with NIST traceability
- Made with 10 Micro coating