Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 13.02 mm, 2 in
Supplier:  MSC 40440471
Encompass
This Class ZZ Minus Plug & Pin Gage is ideal for precision measurement applications. Manufactured from steel, it features a 13.02 mm diameter and a 2 inch overall length, ensuring accuracy for setting micrometers, NoGo inspection, verniers, and other measuring devices. It includes a Certificate of Accuracy with NIST Traceability.
  • Class ZZ Minus tolerance for precise measurements.
  • 13.02 mm diameter suitable for specific gauging requirements.
  • 2 inch overall length for ease of handling.
  • Steel construction for durability.
  • Ideal for setting micrometers and NoGo inspection.
  • Includes certificate of accuracy with NIST traceability.
Catalog No. 50-005-27036