Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 20.53 mm Diameter, 2 in OAL, 10 Micro Coating
Supplier:  MSC 40447989
Encompass
This Class ZZ Minus Plug & Pin Gage features a 20.53 mm diameter and a 2 inch overall length. It is constructed from steel with a 10 Micro coating/finish. This gage is suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices. It also includes a Certificate of Accuracy with NIST Traceability.
  • Has a 20.53 mm diameter
  • Features a 2 inch overall length
  • Constructed from steel
  • Includes a 10 Micro coating/finish
  • Suitable for setting micrometers
  • Ideal for NoGo inspection
  • Can be used with verniers and other measuring devices
  • Comes with a Certificate of Accuracy with NIST Traceability
  • Provides a tolerance of 0.00508 mm
Catalog No. 50-005-28518