Learn More
MSC Vermont Gage Class ZZ Plug Gage, 9.73 mm Diameter, Tool Steel
Supplier: MSC 68100783
This Class ZZ Plus Plug Gage is designed for precise Go inspection and for setting micrometers, verniers, and other measuring devices. Constructed from durable tool steel, it features a 9.73 mm diameter and a 2.0000 inch overall length. It adheres to ASME B89.1 standards and includes a Certificate of Accuracy with NIST Traceability.
- For go inspection and setting measuring devices.
- 9.73 mm diameter.
- 2.0000 inch overall length.
- Constructed from tool steel.
- 0.00508 mm tolerance.
- 10 micro coating/finish.
- Complies with ASME B89.1 standards.
- Includes certificate of accuracy with NIST traceability.