Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.2220 in Diameter, 2.0000 in OAL, Tool Steel
Supplier: MSC 88922208
This Class ZZ Plus Plug Gage is engineered for precise measurement and inspection in demanding environments. With a precise 0.2220 inch diameter and an overall length of 2.0000 inches, it is crafted from durable tool steel. This gage is an essential tool for 'go' inspection, as well as for calibrating micrometers, verniers, and other measuring instruments, guaranteeing high accuracy across various industrial and laboratory applications.
- Precise measurement diameter of 0.2220 in
- Overall length of 2.0000 in
- High tolerance of 0.000200 in
- Constructed from durable tool steel
- Features a 10 micro coating/finish
- Ideal for go inspection and setting measuring devices
- Meets ASME B89.1 standards
- Includes a certificate of accuracy with NIST traceability