Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.62 mm Diameter, 2.0000 inch Length, 10 Micro Finish
Supplier:  MSC 68091677
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement and inspection tasks. Made from durable tool steel, it features a 0.62 mm diameter and a 10 micro finish, ensuring accuracy for applications such as go inspection, setting micrometers, verniers, and other measuring devices. Each gage includes a Certificate of Accuracy with NIST traceability.
  • Overall length of 2.0000 inch
  • Tolerance of 0.00508 mm
  • Constructed from tool steel
  • Ideal for go inspection, setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Features a 10 micro finish
Catalog No. 50-005-27912