Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 6.33 mm Dia, 2.000 in OAL, 0.00508 mm Tolerance
Supplier:  MSC 68097385
Encompass
This Class ZZ Plus Plug Gage features a 6.33 mm diameter and a 2.000 inch overall length. Constructed from durable tool steel, it is designed for precise Go inspection and for accurately setting micrometers, verniers, and various other measuring devices.
  • Used for Go inspection
  • Can be used to set micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Features a 10 micro finish
  • Made from tool steel
  • Has a 0.00508 mm tolerance
Catalog No. 50-005-30342