Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 8.10 mm, 2 in OAL, Tool Steel
Supplier:  MSC 68099159
Encompass
This Class ZZ Plus Plug Gage features an 8.10 mm diameter and a 0.00508 mm tolerance. Constructed from durable tool steel, it is designed for precise 'Go' inspection and for setting micrometers, verniers, and other measuring devices. Each gage includes a Certificate of Accuracy with NIST traceability, ensuring reliable and accurate measurements.
  • Features an 8.10 mm diameter for precise measurement.
  • Provides a 0.00508 mm tolerance for high accuracy.
  • Constructed from durable tool steel.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for 'Go' inspection applications.
  • Ideal for setting micrometers, verniers, and other measuring devices.
  • Meets ASME B89.1 standards.
  • Overall length of 2 in.
Catalog No. 50-005-30669