Learn More
MSC Vermont Gage Pin Gage, 0.3620 Inch Diameter, 2 Inch Length, Class X
Supplier: MSC 88736202
This pin gage is designed for precise measurement applications, suitable for calibrating measuring equipment or directly gaging parts. It offers accurate, cost-effective measurement standards for various industrial settings.
- Class X tolerance
- Go (Plus) tolerance of 0.00004 inch
- NIST traceable
- 4 Micro coating/finish
- Available in sizes every 0.0001 inch up to 1.0000 inch
- Made from tool steel, 60/65 Rc (cold stabilized)
- 2 inch length