Learn More
MSC Vermont Gage Pin Gage, 0.4090 in Diameter, 2 in Length, Class ZZ Minus
Supplier: MSC 50745124
This Class ZZ Minus Pin Gage is designed for precise measurement and inspection applications. Made from durable steel with a black oxide finish, it features a 0.4090 inch diameter and a 2 inch overall length, providing accuracy for various gaging tasks.
- Designed for setting micrometers, verniers, and other measuring devices.
- Ideal for no-go inspection.
- Constructed from steel for durability.
- Features a black oxide coating for enhanced finish.
- Includes a certificate of accuracy with NIST traceability.
- Accurate 0.4090 inch diameter with a 0.000200 inch tolerance.
- Overall length of 2 "