Learn More
MSC Vermont Gage Pin Gage, 0.4815 inch, Class ZZ, Go (Plus), Black Guard
Supplier: MSC 50758770
This economical general purpose pin gage provides quick and accurate measurements for various applications. It is ideal for measuring hole sizes, calculating distances between holes, gaging slot widths, checking locations, and for Go/NoGo gaging. This versatile tool can also be used to size in micrometers and snap gages.
- Made from tool steel with a hardness of 60/65 Rc.
- Features Class ZZ accuracy with a .0002 inch or .005 mm tolerance.
- NIST traceable for reliable measurements.
- "Go (Plus)" indicates a unilateral tolerance on the plus side.
- Round within .0001 inch or .0025 mm.
- Measures 2 inches long.
- Offers a 10 microinch finish or better.
- Gages over .0605 inches are marked with size and direction of tolerance.
- Includes a certificate of accuracy.
- Each pin is individually packaged.
- Provides added wear life compared to closer tolerance gages.