Learn More
MSC Vermont Gage Pin Gage, 10.66 mm, Class ZZ, Minus Tolerance
Supplier:  MSC 40438111
Encompass
This pin gage is designed for precise measurement of hole sizes, featuring a 10.66 mm diameter and a Class ZZ minus tolerance of +0.000/-0.005 inches. Manufactured from durable tool steel, it has an overall length of 2 inches and a 10 microfinish or better, ensuring high accuracy for inspection and setting measuring devices.
  • Made from durable tool steel.
  • Features a 10 microfinish or better for high accuracy.
  • Includes a certificate of accuracy with NIST traceability.
  • Ideal for no-go inspection and setting micrometers, verniers, and other measuring devices.
Catalog No. 50-005-27034