Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.0470 in Dia, 2 in Long
Supplier:  MSC 88704705
Encompass
This pin gage is a Class X Plus precision tool designed for accurate measurement and calibration. It features a 0.0470 inch diameter and is 2 inches long, making it suitable for a variety of applications requiring high precision.
  • 4 micro coated finish
  • Constructed from durable tool steel
  • Ideal for calibration of measuring equipment or gaging parts directly
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-31075