Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.1565 in Dia, 2 in Long
Supplier:  MSC 88715651
Encompass
This Class X Plus Pin Gage features a 0.1565 inch diameter and a 2 inch length. It is designed for the calibration of measuring equipment or for direct gaging of parts, featuring a 4 Micro coating for enhanced performance.
  • Includes certificate of accuracy with NIST traceability
  • Calibrates measuring equipment or gages parts directly
  • Features a 4 Micro coating
Catalog No. 50-005-31080