Learn More
MSC Vermont Gage Pin Gage, Class ZZ, 0.453 in Diameter, Tool Steel
Supplier: MSC 88945308
This pin gage is crafted from tool steel with a Rockwell hardness of 60/65 and features a steel finish with 10 microfinish or better. It has a diameter of 0.4530 and a tolerance of +.0002/-.0000. With an overall length of 2 inches, this go (plus) type pin gage is designed to accurately measure hole sizes and includes a certificate of accuracy with NIST traceability.
- Constructed from tool steel with Rc 60/65 hardness
- Features a tolerance of +.0002/-.0000
- Designed to measure hole sizes within +.0002/-.0000
- Includes a certificate of accuracy with NIST traceability
- Overall length of 2 in
- Diameter of 0.4530
- Class ZZ
- Steel finish with 10 microfinish or better
- Go (plus) type pin gage