Learn More
MSC Vermont Gage Pin Gage, Class ZZ Minus, 13.62 mm, Tool Steel
Supplier:  MSC 40441073
Encompass
This pin gage is designed for precise measurement of hole sizes. It features a 13.62 mm diameter and is constructed from durable tool steel. The gage is Class ZZ Minus with a tolerance of +.000/-.005, ensuring high accuracy for various applications.
  • Constructed from tool steel with Rc 60/65 hardness
  • Steel finish with 10 microfinish or better
  • Overall length is 2 in
  • Measures hole sizes within +.000/-.005"
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-28901