Learn More
MSC Vermont Gage Pin Gage, Class ZZ Plus, 14.75 mm, Tool Steel
Supplier:  MSC 68105808
Encompass
This pin gage is designed for precise measurement and inspection applications. It features a specific diameter and tolerance, making it suitable for verifying dimensions and calibrating various measuring instruments.
  • Class ZZ Plus
  • 14.75 mm diameter
  • Plus 0.00508 mm tolerance
  • Made from durable tool steel
  • 10 microfinish for accuracy
  • Reversible style
  • Includes a Certificate of Accuracy with NIST Traceability
  • Conforms to ASME B89.1 standards
  • Ideal for go inspection, setting micrometers, verniers, and other measuring devices
Catalog No. 50-005-31409