Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Plus, Steel, 10.55 mm, Metric
Supplier:  MSC 68101609
Encompass
This Vermont Gage pin gage is a Class ZZ, plus tolerance, steel gage with a 10.55 mm size. Designed for precise measurements in metric systems, it ensures accurate checking of hole sizes.
  • Pin gage
  • Class Zz
  • Plus direction of tolerance
  • Steel material
  • 10.55 mm size
  • Reversible style
  • Metric system of measurement
Catalog No. 50-005-31405