Learn More
MSC Vermont Gage Pin Gage, Minus, Class ZZ, 0.8690 Inch Diameter
Supplier: MSC 89086904
This pin gage is designed for precise measurement of hole sizes. It features a minus tolerance, Class ZZ accuracy, and an overall length of 2 inches, making it suitable for various industrial and laboratory applications.
- Constructed from tool steel with a hardness of Rc 60/65.
- Features a tolerance of +.0000/-.0002.
- Ideal for measuring hole sizes with high precision.
- Includes a certificate of accuracy with NIST traceability.
- Overall length is 2 inches.
- Designed as a NoGo (minus) type gage.
- Finish is steel with 10 microfinish or better.
- Class ZZ for high accuracy.