Learn More
MSC Vermont Gage Pin Gage, Minus, Class ZZ, 17.90 mm
Supplier: MSC 40445355
This pin gage is constructed from tool steel with an Rc 60/65 hardness and features a steel finish with 10 microfinish or better. It has a diameter of 17.90 mm and an overall length of 2 in. This gage is designed for precise measurements with a tolerance of +0.000/-0.005 in.
- Tool steel construction with Rc 60/65 hardness
- Precision tolerance of +0.000/-0.005 in
- Measures hole sizes accurately
- Includes a certificate of accuracy with NIST traceability
- Overall length of 2 in
- NoGo (Minus) type for specific gaging
- Steel finish with 10 microfinish or better
- Class ZZ designation for accuracy