Learn More
MSC Vermont Gage Pin Gage, Minus, Class ZZ, 7.23mm, 2 in OAL
Supplier:  MSC 40434680
Encompass
This pin gage is designed for precise measurement of hole sizes. It features a tool steel construction with a Rc 60/65 hardness and a 10 microfinish or better. The gage has a 7.23mm diameter, a 2 in overall length, and a tight tolerance of +.000/-.005.
  • Measures hole sizes within +.000/-.005"
  • Includes certificate of accuracy with NIST traceability
  • NoGo (minus) type
  • Class ZZ
  • Made from tool steel with Rc 60/65 hardness
  • Steel finish with 10 microfinish or better
Catalog No. 50-005-27027