Learn More
MSC Vermont Gage Plug Gage, 0.0610 inch Diameter, Class ZZ Plus, Tool Steel
Supplier: MSC 88906102
The Vermont Gage Class ZZ Plus Plug Gage is a precision measuring tool designed for accurate "Go" inspection and for calibrating other measuring devices. Constructed from durable tool steel with a 10 micro finish, it ensures reliable performance in various industrial and metrology applications.
- Designed for "Go" inspection.
- Suitable for setting micrometers and verniers.
- Made from durable tool steel.
- Features a 10 micro finish.
- Includes a certificate of accuracy with NIST traceability.
- Conforms to ASME B89.1 standards.
- Precise 0.0610 inch diameter.
- Overall length of 2.0000 inches.
- Tight tolerance of 0.000200 inches.