Learn More
MSC Vermont Gage Plug Gage, 0.2710 inch Diameter, ZZ Plus Class, Tool Steel
Supplier:  MSC 50743798
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is a precision measuring instrument with a 0.2710 inch diameter and an overall length of 2.0000 " It features a tight tolerance of 0.000200 inch, making it suitable for accurate "Go" inspection applications. Constructed from durable tool steel and finished with black oxide, this plug gage is designed for setting micrometers, verniers, and various other measuring devices.
  • Precision measuring instrument for accurate "Go" inspection applications
  • Diameter of 0.2710 inch
  • Overall length of 2.0000 inch
  • Tolerance of 0.000200 inch
  • Constructed from durable tool steel
  • Features a black oxide finish
  • Includes a certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Suitable for setting micrometers, verniers, and other measuring devices
Catalog No. 50-005-28192