Learn More
MSC Vermont Gage Plug Gage, 0.5003 Inch, Class X, No Go
Supplier:  MSC 80346588
Encompass
This precision steel plug gage is designed for checking hole dimensions, calibrating equipment, or gaging parts directly. It features a 0.5003 inch diameter with a Class X tolerance and a No Go type, ensuring accurate checks for oversized parts.
  • Accurately checks hole dimensions and calibrates measuring equipment.
  • Features a 0.5003 inch diameter with 0.00004 inch accuracy and Class X tolerance.
  • No Go minus gage type prevents oversized parts.
  • Constructed from durable steel with an uncoated finish; 2 inch overall length.
  • Surface finish is four microinch or better.
  • Includes a certificate of accuracy with NIST traceability and meets ISO 17025 standard.
Catalog No. 50-005-30401