Learn More
MSC Vermont Gage Plug Gage, 15.65 mm, 0.00508 mm Tolerance, Tool Steel
Supplier:  MSC 68106707
Encompass
This plug gage is designed for "Go Inspection" and for setting micrometers, verniers, and other measuring devices. It features a 15.65 mm diameter and is constructed from tool steel, with a 0.00508 mm tolerance and a 10 Micro coating/finish. This product includes a Certificate of Accuracy with NIST Traceability and adheres to ASME B89.1 Standards.
  • Designed for "Go Inspection"
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Tolerance of 0.00508 mm
  • Features a 10 micro coating/finish
  • Includes a certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards
  • Constructed from tool steel
  • Country of origin is the United States
Catalog No. 50-005-27962