Learn More
MSC Vermont Gage Plug Gage, 8.18 mm Diameter, 0.00508 mm Tolerance, Tool Steel
Supplier:  MSC 68099233
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is designed for "Go Inspection" and can be used to set micrometers, verniers, and other measuring devices. It includes a Certificate of Accuracy with NIST Traceability, ensuring reliable and precise measurements. Made from durable tool steel, this gage offers a 0.00508 mm tolerance and a 10 Micro finish for high accuracy and longevity.
  • Designed for go inspection
  • Can be used to set micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Made from durable tool steel
  • Offers a 0.00508 mm tolerance
  • Features a 10 micro finish
  • Manufactured in the United States
  • 8.18 mm diameter
  • 2.0000 in overall length
  • Meets ASME B89.1 standards
Catalog No. 50-005-28065