Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.2960 in Dia, 2.0000 in OAL, Tool Steel
Supplier:  MSC 88929609
Encompass
This plug gage is an essential tool for precise dimensional verification and calibration tasks. It is ideal for 'Go' inspection, as well as accurately setting micrometers, verniers, and other measuring instruments. Constructed from durable tool steel, this gage ensures reliable performance and accuracy in demanding environments.
  • Designed for go inspection.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Constructed from tool steel.
  • Features a 10 micro finish.
  • Conforms to ASME B89.1 standards.
  • Provides a tolerance of 0.000200 in.
Catalog No. 50-005-30536